Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006930.3 | NP_008861.2 | 3 | 305 | ||
NM_170679.2 | NP_733779.1 | 3 | 305 | |||
GenBank mRNA | BC009839.2 | - | 3 | 189 | 4053593 | |
BC020798.1 | - | 3 | 240 | 4243711 | ||
BC025673.1 | - | 3 | 202 | 5203008 | ||
BC065730.1 | - | 3 | 226 | 6672613 | ||
BI552966.1 | - | 3 | 272 | |||
BI670085.1 | - | 3 | 310 | |||
U33760.1 | - | 3 | 219 | |||
U37558.1 | - | 2 | 87 | |||
Z47087.1 | - | 3 | 235 |