Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLECL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253750.1 | NP_001240679.1 | 1 | 171 | ||
NM_001267701.1 | NP_001254630.1 | 1 | 171 | |||
NM_172004.3 | NP_742001.1 | 1 | 171 | |||
XM_011520574.2 | XP_011518876.1 | 1 | 214 | |||
GenBank mRNA | AF518873.1 | - | 1 | 136 | ||
BC093859.1 | - | 1 | 171 | 7939704 |