Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153605.3 | NP_705833.3 | 12 | 7781 | ||
XM_005247117.4 | XP_005247174.1 | 11 | 7484 | |||
GenBank mRNA | AK091199.1 | - | 4 | 252 | ||
BX647079.1 | - | 4 | 269 | |||
BX648353.1 | - | 9 | 1831 | |||
CR749313.1 | - | 9 | 906 |