Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PANK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324191.1 | NP_001311120.1 | 3 | 1156 | ||
NM_001324192.1 | NP_001311121.1 | 2 | 988 | |||
NM_001324193.1 | NP_001311122.1 | 2 | 324 | |||
NM_024960.5 | NP_079236.3 | 2 | 259 | |||
NM_153638.3 | NP_705902.2 | 2 | 988 | |||
NM_153640.3 | NP_705904.1 | 2 | 324 | |||
NR_136715.1 | 2 | 988 | ||||
XM_005260835.3 | XP_005260892.1 | 2 | 441 | |||
XM_005260836.4 | XP_005260893.3 | 2 | 261 | |||
XM_011529364.2 | XP_011527666.1 | 2 | 844 | |||
XM_011529365.2 | XP_011527667.1 | 2 | 844 | |||
XM_017028077.1 | XP_016883566.1 | 2 | 361 | |||
XM_017028078.1 | XP_016883567.1 | 2 | 1702 | |||
XM_017028079.1 | XP_016883568.1 | 2 | 272 | |||
GenBank mRNA | AF494409.1 | - | 2 | 827 | ||
AK021791.1 | - | 2 | 259 | |||
AK097796.1 | - | 2 | 507 | |||
AK310190.1 | - | 3 | 696 | |||
AL519408.3 | - | 2 | 276 | |||
AL532307.3 | - | 2 | 332 | |||
BC009421.1 | - | 2 | 504 | |||
BC065019.1 | - | 2 | 270 | |||
BC107724.1 | - | 2 | 311 | |||
BF698882.1 | - | 308 |