Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PODN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153703.4 | NP_714914.2 | 9, 10 | 1981 | ||
XM_005270457.3 | XP_005270514.1 | 10, 11 | 2010 | |||
XM_011540677.2 | XP_011538979.1 | 5, 6 | 1135 | |||
XM_017000294.1 | XP_016855783.1 | 9, 10 | 2331 | |||
GenBank mRNA | AK296649.1 | - | 10, 11 | 1876 | ||
AK299375.1 | - | 11, 12 | 2066 | |||
AK300847.1 | - | 4, 5 | 921 | |||
AK304599.1 | - | 5, 6 | 1394 | |||
AK314096.1 | - | 9, 10 | 1575 | |||
AY313607.1 | - | 9, 10 | 1820 | |||
AY358294.1 | - | 11, 12 | 2097 | |||
BC028292.1 | - | 9, 10 | 1804 | 4938380 | ||
BC030608.2 | - | 9, 10 | 1822 | 4826932 | ||
BQ719232.1 | - | 2, 3 | 643 |