Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153704.5 | NP_714915.3 | 2 | 326 | ||
NR_024522.1 | 2 | 326 | ||||
XM_011517363.2 | XP_011515665.1 | 2 | 298 | |||
GenBank mRNA | AK094935.1 | - | 2 | 250 | ||
AK308071.1 | - | 3 | 216 | |||
AK308735.1 | - | 2 | 275 | |||
BC032835.2 | - | 2 | 244 | 4825770 | ||
BC054338.1 | - | 2 | 253 | |||
BX648768.1 | - | 2 | 270 | |||
DA928293.1 | - | 2 | 305 | |||
DC368894.1 | - | 3 | 216 | |||
KU178827.1 | - | 2 | 225 | |||
KU178828.1 | - | 2 | 225 | |||
KU178829.1 | - | 2 | 225 |