Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GALT5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026542.1 | 3 | 670 | |||
GenBank mRNA | AF426266.1 | - | 3 | 289 | ||
AK129590.1 | - | 2 | 377 | |||
BC119737.2 | - | 3 | 263 | |||
BC119738.2 | - | 3 | 263 |