Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153812.2 | NP_722519.2 | 2, 3 | 517 | ||
XM_011540762.1 | XP_011539064.1 | 2, 3 | 319 | |||
GenBank mRNA | AB209544.1 | - | 2, 3 | 209 | ||
AK027492.1 | - | 1, 2 | 42 | |||
AK097715.1 | - | 2, 3 | 451 | |||
AK315110.1 | - | 2, 3 | 449 | |||
AY069975.1 | - | 2, 3 | 147 | |||
BC038516.1 | - | 2, 3 | 528 | 5266750 |