Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004841.3 | NP_004832.1 | 3 | 278 | ||
NM_170692.2 | NP_733793.2 | 5 | 1361 | |||
XM_005245622.3 | XP_005245679.1 | 3 | 934 | |||
XM_011510166.2 | XP_011508468.1 | 5 | 1361 | |||
XM_011510167.2 | XP_011508469.1 | 5 | 1361 | |||
XM_017002849.1 | XP_016858338.1 | 5 | 3967 | |||
XM_017002850.1 | XP_016858339.1 | 5 | 3966 | |||
XM_017002851.1 | XP_016858340.1 | 5 | 3966 | |||
XM_017002852.1 | XP_016858341.1 | 5 | 3965 | |||
XM_017002853.1 | XP_016858342.1 | 5 | 1405 | |||
XM_017002854.1 | XP_016858343.1 | 3 | 937 | |||
XM_017002855.1 | XP_016858344.1 | 3 | 936 | |||
GenBank mRNA | AB209789.1 | - | 5 | 1269 | ||
AF047711.1 | - | 3 | 278 | |||
AK075169.1 | - | 5 | 1361 | |||
AK290308.1 | - | 3 | 221 | |||
AK311017.1 | - | 5 | 1360 | |||
AV706305.1 | - | 5 | 647 | |||
BC041045.1 | - | 914 | ||||
BC063442.1 | - | 5 | 921 | |||
BC078167.1 | - | 5 | 983 | |||
BC110611.2 | - | 5 | 593 | 40028914 |