Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLECL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253750.1 | NP_001240679.1 | 2 | 412 | ||
NM_001267701.1 | NP_001254630.1 | 2 | 412 | |||
NM_172004.3 | NP_742001.1 | 2 | 412 | |||
XM_011520574.2 | XP_011518876.1 | 2 | 455 | |||
XM_017018885.1 | XP_016874374.1 | 3 | 292 | |||
GenBank mRNA | AF518873.1 | - | 2 | 377 | ||
BC093857.1 | - | 2 | 412 | 7939702 | ||
BC093859.1 | - | 2 | 412 | 7939704 | ||
BX103210.1 | - | 2 | 185 | |||
DA619574.1 | - | 2 | 237 |