Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301062.1 | NP_001287991.1 | 4 | 1427 | ||
NM_153608.2 | NP_705836.1 | 6 | 1762 | |||
XM_011526574.2 | XP_011524876.1 | 6 | 1676 | |||
XM_017026414.1 | XP_016881903.1 | 6 | 1648 | |||
XM_017026415.1 | XP_016881904.1 | 5 | 1614 | |||
GenBank mRNA | AK075062.1 | - | 5 | 1489 | ||
AK291575.1 | - | 6 | 1832 | |||
BC014935.2 | - | 6 | 1771 | |||
BC034032.1 | - | 1 | 811 | |||
BC125068.1 | - | 1 | 1488 | 40117544 | ||
BC125069.1 | - | 4 | 1357 | 40117550 | ||
BU182389.1 | - | 1 | 13 |