Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153615.1 | NP_705843.1 | 3 | 1674 | ||
GenBank mRNA | AK131340.1 | - | 5 | 2112 | ||
AY101396.1 | - | 3 | 1674 | |||
BC101108.3 | - | 2 | 1354 | 40011557 | ||
BC101109.2 | - | 2 | 1354 | 40011564 | ||
BC101110.3 | - | 3 | 1229 |