Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDELC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153705.4 | NP_714916.3 | 5 | 1098 | ||
XM_011542621.2 | XP_011540923.1 | 5 | 1071 | |||
GenBank mRNA | AF533708.1 | - | 5 | 1057 | ||
AK098206.1 | - | 1 | 63 | |||
AK125519.1 | - | 4 | 880 | |||
AK313534.1 | - | 5 | 960 | |||
AY358616.1 | - | 4 | 1167 | |||
BC036526.2 | - | 5 | 1052 | 5266665 |