Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153812.2 | NP_722519.2 | 4 | 3179 | ||
XM_011540762.1 | XP_011539064.1 | 4 | 2981 | |||
GenBank mRNA | AL121733.1 | - | 2 | 2667 | ||
AY069975.1 | - | 4 | 2809 | |||
BC038516.1 | - | 4 | 3189 | 5266750 |