Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170710.4 | NP_733828.2 | 30 | 4071 | ||
NM_181265.3 | NP_851782.3 | 28 | 3903 | |||
XM_005262725.3 | XP_005262782.1 | 28 | 3570 | |||
XM_011531582.2 | XP_011529884.1 | 29 | 3912 | |||
XM_017007686.1 | XP_016863175.1 | 29 | 3900 | |||
XM_017007687.1 | XP_016863176.1 | 30 | 3985 | |||
XM_017007688.1 | XP_016863177.1 | 30 | 3902 | |||
XM_017007689.1 | XP_016863178.1 | 28 | 3878 | |||
XM_017007690.1 | XP_016863179.1 | 29 | 3964 | |||
XM_017007691.1 | XP_016863180.1 | 28 | 3734 | |||
XM_017007692.1 | XP_016863181.1 | 29 | 3820 | |||
XM_017007693.1 | XP_016863182.1 | 27 | 3713 | |||
XM_017007694.1 | XP_016863183.1 | 28 | 3697 | |||
XM_017007695.1 | XP_016863184.1 | 28 | 3748 | |||
XM_017007696.1 | XP_016863185.1 | 27 | 3727 | |||
GenBank mRNA | AF492460.1 | - | 30 | 3924 | ||
AK130128.1 | - | 14 | 1533 |