Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170776.4 | NP_740746.4 | 3 | 351 | ||
XM_005255842.3 | XP_005255899.1 | 3 | 404 | |||
XM_006721170.3 | XP_006721233.1 | 3 | 404 | |||
XM_011522953.2 | XP_011521255.1 | 3 | 404 | |||
XM_011522954.1 | XP_011521256.1 | 3 | 404 | |||
GenBank mRNA | AB049169.1 | - | 3 | 294 | ||
AK172801.1 | - | 3 | 351 | |||
AK310124.1 | - | 3 | 290 | |||
AY140959.1 | - | 3 | 248 | |||
BC064508.1 | - | 3 | 337 | 6424664 |
Set Membership: |
G-protein coupled receptor |