Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308217.1 | NP_001295146.1 | 2 | 364 | ||
NM_001308222.1 | NP_001295151.1 | 2 | 1117 | |||
NM_003471.3 | NP_003462.2 | 2 | 484 | |||
NM_172159.3 | NP_751891.1 | 2 | 1167 | |||
NM_172160.2 | NP_751892.1 | 2 | 364 | |||
XM_011513115.2 | XP_011511417.1 | 2 | 77 | |||
XM_017007171.1 | XP_016862660.1 | 2 | 154 | |||
XM_017007172.1 | XP_016862661.1 | 2 | 1370 | |||
XM_017007174.1 | XP_016862663.1 | 2 | 350 | |||
GenBank mRNA | AK057059.1 | - | 2 | 900 | ||
AK127240.1 | - | 2 | 1097 | |||
AK292693.1 | - | 2 | 350 | |||
AK292999.1 | - | 2 | 416 | |||
AK303287.1 | - | 2 | 334 | |||
AY780786.1 | - | 2 | 246 | |||
BC043166.2 | - | 2 | 1117 | 5286833 | ||
DA408045.1 | - | 2 | 462 | |||
DC410397.1 | - | 2 | 364 | |||
L39833.1 | - | 2 | 355 | |||
L47665.1 | - | 2 | 353 | |||
U16953.1 | - | 2 | 334 | |||
U17968.1 | - | 2 | 259 | |||
U33428.1 | - | 2 | 273 |