Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002482.3 | NP_002473.2 | 3 | 368 | ||
NM_152298.3 | NP_689511.2 | 3 | 368 | |||
XM_005270888.2 | XP_005270945.1 | 4 | 388 | |||
XM_005270889.2 | XP_005270946.1 | 4 | 388 | |||
XM_011541509.1 | XP_011539811.1 | 4 | 388 | |||
XM_017001356.1 | XP_016856845.1 | 3 | 315 | |||
GenBank mRNA | AF035191.1 | - | 3 | 293 | ||
AK056161.1 | - | 4 | 384 | |||
AK222778.1 | - | 3 | 311 | |||
AK222816.1 | - | 3 | 311 | |||
AK291637.1 | - | 3 | 311 | |||
AY700118.1 | - | 1 | 296 | |||
BC003113.1 | - | 3 | 286 | 3504220 | ||
BC009933.2 | - | 3 | 292 | |||
BC010105.2 | - | 3 | 296 | 3538429 | ||
BC011580.1 | - | 3 | 294 | 4558913 | ||
BC053608.1 | - | 3 | 291 | 6013263 | ||
BT006757.1 | - | 3 | 209 | |||
DC361634.1 | - | 3 | 368 | |||
M97856.1 | - | 3 | 294 |