Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172193.2 | NP_751943.1 | 3 | 354 | ||
XM_011536422.2 | XP_011534724.1 | 3 | 277 | |||
GenBank mRNA | AF111806.1 | - | 3 | 292 | ||
AK127202.1 | - | 3 | 277 | |||
BC031270.1 | - | 3 | 317 | |||
BC101595.1 | - | 3 | 354 | 8069101 | ||
BC101597.1 | - | 3 | 354 | 8069103 |