Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114101.1 | NP_001107573.1 | 3 | 467 | ||
NM_172369.3 | NP_758957.2 | 3 | 444 | |||
GenBank mRNA | AF087892.1 | - | 3 | 442 | ||
AK057792.1 | - | 3 | 459 | |||
AK130613.1 | - | 3 | 442 | |||
AK226152.1 | - | 1 | 3739 | |||
BC009016.1 | - | 3 | 409 | 4212772 | ||
CB995661.1 | - | 3 | 494 | |||
DA849505.1 | - | 3 | 436 |