Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCD2OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164839.1 | NP_001158311.1 | 2 | 395 | ||
NM_173472.1 | NP_775743.1 | 2 | 395 | |||
XM_011533330.2 | XP_011531632.1 | 3 | 863 | |||
XM_011533331.2 | XP_011531633.1 | 2 | 538 | |||
GenBank mRNA | AF230334.1 | - | 2 | 387 | ||
AY248899.1 | - | 2 | 245 | |||
BC028122.1 | - | 2 | 395 | 5168387 |