Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172128.1 | NP_001165599.1 | 14 | 2888 | ||
NM_004513.5 | NP_004504.3 | 2 | 640 | |||
NM_172217.3 | NP_757366.2 | 13 | 2628 | |||
XM_005254342.3 | XP_005254399.1 | 14 | 2925 | |||
XM_005254346.4 | XP_005254403.1 | 2 | 2058 | |||
XM_011521519.2 | XP_011519821.1 | 13 | 2628 | |||
XM_011521520.2 | XP_011519822.1 | 14 | 2903 | |||
XM_017022140.1 | XP_016877629.1 | 13 | 2628 | |||
XM_017022141.1 | XP_016877630.1 | 14 | 2887 | |||
XM_017022142.1 | XP_016877631.1 | 14 | 2903 | |||
XM_017022143.1 | XP_016877632.1 | 13 | 2776 | |||
XM_017022144.1 | XP_016877633.1 | 14 | 2941 | |||
XM_017022145.1 | XP_016877634.1 | 14 | 2644 | |||
XM_017022146.1 | XP_016877635.1 | 4 | 1210 | |||
GenBank mRNA | AK124565.1 | - | 12 | 2230 | ||
AK126222.1 | - | 3 | 1409 | |||
AK128740.1 | - | 8 | 2497 | |||
AK131530.1 | - | 14 | 2887 | |||
AK222512.1 | - | 2 | 565 | |||
AK303659.1 | - | 4 | 1249 | |||
AL833554.1 | - | 16 | 2656 | |||
AY324389.1 | - | 13 | 2527 | |||
BC050362.1 | - | 2 | 600 | |||
BC064911.1 | - | 2 | 525 | |||
BC136660.1 | - | 14 | 2653 | 9020650 | ||
CR749286.1 | - | 15 | 3348 | |||
FJ032370.1 | - | 1 | 438 | |||
M90391.1 | - | 3 | 727 | |||
U82972.1 | - | 2 | 604 |