Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001551.2 | NP_001542.1 | 2 | 459 | ||
XM_017029489.1 | XP_016884978.1 | 1 | 660 | |||
XM_017029490.1 | XP_016884979.1 | 1 | 660 | |||
GenBank mRNA | AK054596.1 | - | 1 | 660 | ||
AK315660.1 | - | 1 | 577 | |||
BC004137.2 | - | 2 | 459 | 2962967 | ||
BT006736.1 | - | 1 | 161 | |||
CR456782.1 | - | 1 | 161 | |||
Y08915.1 | - | 1 | 169 |