Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321381.1 | NP_001308310.1 | 5, 6 | 993 | ||
NM_173079.3 | NP_775102.2 | 4, 5 | 987 | |||
GenBank mRNA | AL831813.1 | - | 4, 5 | 970 | ||
AY189284.1 | - | 5, 6 | 969 | |||
BC014873.1 | - | 4, 5 | 975 | |||
BC039247.1 | - | 4, 5 | 984 | 5271271 |