Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099783.1 | NP_001093253.1 | 2 | 185 | ||
NM_173487.2 | NP_775758.2 | 2 | 457 | |||
XM_005262734.4 | XP_005262791.1 | 2 | 452 | |||
XM_017007722.1 | XP_016863211.1 | 2 | 1514 | |||
XM_017007723.1 | XP_016863212.1 | 2 | 1264 | |||
XM_017007724.1 | XP_016863213.1 | 2 | 440 | |||
GenBank mRNA | AK091022.1 | - | 2 | 446 | ||
AK309229.1 | - | 2 | 431 | |||
BC032582.1 | - | 2 | 388 | 5494700 | ||
BE566349.1 | - | 2 | 140 | |||
BP871612.1 | - | 2 | 457 | |||
BQ549846.1 | - | 2 | 216 | |||
BX538164.1 | - | 2 | 398 | |||
CN483524.1 | - | 2 | 185 |