Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB053324.1 | - | 1957 | |||
AB053325.1 | - | 1946 | ||||
AB053326.1 | - | 1911 |