Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EYA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005244.4 | NP_005235.3 | 16 | 2679 | ||
NM_172110.3 | NP_742108.2 | 14 | 2442 | |||
XM_005260327.2 | XP_005260384.1 | 14 | 2126 | |||
XM_017027721.1 | XP_016883210.1 | 15 | 2307 | |||
GenBank mRNA | AF055015.1 | - | 12 | 1993 | ||
AK093582.1 | - | 7 | 2563 | |||
BC000289.2 | - | 10 | 1831 | |||
BC008803.2 | - | 16 | 2361 | 3944547 | ||
BC013882.2 | - | 14 | 2442 | |||
U71207.1 | - | 15 | 2306 |