Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308217.1 | NP_001295146.1 | 13 | 1663 | ||
NM_001308222.1 | NP_001295151.1 | 13 | 2416 | |||
NM_003471.3 | NP_003462.2 | 14 | 1870 | |||
NM_172159.3 | NP_751891.1 | 14 | 2553 | |||
NM_172160.2 | NP_751892.1 | 14 | 1750 | |||
XM_011513115.2 | XP_011511417.1 | 14 | 1463 | |||
XM_011513116.2 | XP_011511418.1 | 12 | 1542 | |||
XM_011513117.2 | XP_011511419.1 | 12 | 1353 | |||
XM_017007171.1 | XP_016862660.1 | 14 | 1540 | |||
XM_017007172.1 | XP_016862661.1 | 14 | 2756 | |||
XM_017007173.1 | XP_016862662.1 | 12 | 1457 | |||
GenBank mRNA | AK057059.1 | - | 14 | 2286 | ||
AK127240.1 | - | 14 | 2483 | |||
AK296728.1 | - | 12 | 1454 | |||
AK303287.1 | - | 13 | 1633 | |||
BC043166.2 | - | 14 | 2503 | 5286833 | ||
L39833.1 | - | 14 | 1742 | |||
L47665.1 | - | 14 | 1739 | |||
U33428.1 | - | 14 | 1660 |