Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172128.1 | NP_001165599.1 | 19 | 4535 | ||
NM_004513.5 | NP_004504.3 | 7 | 2290 | |||
NM_172217.3 | NP_757366.2 | 18 | 4278 | |||
XM_005254342.3 | XP_005254399.1 | 19 | 4575 | |||
XM_005254346.4 | XP_005254403.1 | 7 | 3708 | |||
XM_011521520.2 | XP_011519822.1 | 19 | 4553 | |||
XM_017022140.1 | XP_016877629.1 | 18 | 4772 | |||
XM_017022141.1 | XP_016877630.1 | 19 | 4537 | |||
XM_017022142.1 | XP_016877631.1 | 19 | 4550 | |||
XM_017022143.1 | XP_016877632.1 | 18 | 4426 | |||
XM_017022144.1 | XP_016877633.1 | 19 | 4591 | |||
XM_017022146.1 | XP_016877635.1 | 9 | 2860 | |||
GenBank mRNA | AK124565.1 | - | 17 | 3877 | ||
AK126222.1 | - | 8 | 3056 | |||
AK222512.1 | - | 7 | 2212 | |||
CR749286.1 | - | 20 | 4998 | |||
M90391.1 | - | 8 | 2377 |