Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEIS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001220482.1 | NP_001207411.1 | 13 | 3062 | ||
NM_002399.3 | NP_002390.1 | 13 | 3013 | |||
NM_170674.4 | NP_733774.1 | 13 | 3291 | |||
NM_170675.4 | NP_733775.1 | 12 | 3216 | |||
NM_170676.4 | NP_733776.1 | 12 | 3195 | |||
NM_170677.4 | NP_733777.1 | 13 | 3312 | |||
NM_172315.2 | NP_758526.1 | 12 | 2788 | |||
NM_172316.2 | NP_758527.1 | 13 | 3078 | |||
NR_051953.1 | 12 | 3214 | ||||
XM_006720523.1 | XP_006720586.1 | 12 | 2606 | |||
XM_006720524.1 | XP_006720587.1 | 13 | 2702 | |||
XM_006720525.1 | XP_006720588.1 | 13 | 2681 | |||
XM_006720526.2 | XP_006720589.1 | 12 | 3602 | |||
XM_006720527.2 | XP_006720590.1 | 11 | 3483 | |||
XM_006720529.2 | XP_006720592.1 | 12 | 3579 | |||
XM_011521591.2 | XP_011519893.1 | 10 | 2668 | |||
XM_017022205.1 | XP_016877694.1 | 13 | 3677 | |||
GenBank mRNA | AF017418.1 | - | 6 | 1805 | ||
AK001298.1 | - | 3 | 1896 | |||
AK226072.1 | - | 13 | 3315 | |||
BC001516.2 | - | 13 | 3015 | 2964406 | ||
BC001844.2 | - | 13 | 3015 | 2964406 | ||
BC007202.1 | - | 13 | 3015 | 2964406 |