Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001719.2 | NP_001710.1 | 2, 3 | 1148 | ||
GenBank mRNA | AK291186.1 | - | 2, 3 | 724 | ||
AK312419.1 | - | 2, 3 | 721 | |||
BC008584.1 | - | 2, 3 | 660 | 4183402 | ||
EU679366.1 | - | 2, 3 | 532 | |||
M60316.1 | - | 2, 3 | 715 | |||
X51801.1 | - | 2, 3 | 741 |