Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286075.1 | NP_001273004.1 | 8, 9 | 1023 | ||
NM_004320.4 | NP_004311.1 | 10, 11 | 1374 | |||
NM_173201.3 | NP_775293.1 | 10, 11 | 1374 | |||
GenBank mRNA | AK128456.1 | - | 8, 9 | 1013 | ||
AK291314.1 | - | 10, 11 | 1303 | |||
BC037354.1 | - | 10, 11 | 1302 | |||
BX537389.1 | - | 10, 11 | 1370 | |||
BX647365.1 | - | 10, 11 | 1370 | |||
BX647366.1 | - | 10, 11 | 1366 | |||
BX647367.1 | - | 10, 11 | 1374 | |||
BX647368.1 | - | 3, 4 | 412 | |||
BX648544.1 | - | 10, 11 | 1371 | |||
BX649072.1 | - | 10, 11 | 1363 |