Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278682.1 | NP_001265611.1 | 3 | 536 | ||
NM_001278684.1 | NP_001265613.1 | 4 | 893 | |||
NM_001278686.1 | NP_001265615.1 | 4 | 559 | |||
NM_003244.3 | NP_003235.1 | 3 | 626 | |||
NM_170695.3 | NP_733796.2 | 3 | 1270 | |||
NM_173207.2 | NP_775299.1 | 3 | 504 | |||
NM_173208.2 | NP_775300.1 | 4 | 896 | |||
NM_173209.2 | NP_775301.1 | 3 | 712 | |||
NM_173210.2 | NP_775302.1 | 3 | 663 | |||
NM_173211.1 | NP_775303.1 | 3 | 375 | |||
NM_174886.2 | NP_777480.1 | 4 | 693 | |||
XM_011525735.2 | XP_011524037.1 | 3 | 872 | |||
XM_017025958.1 | XP_016881447.1 | 4 | 957 | |||
XM_017025959.1 | XP_016881448.1 | 3 | 742 | |||
GenBank mRNA | AF179900.1 | - | 3 | 314 | ||
AI813434.1 | - | 4 | 514 | |||
AK026766.1 | - | 2 | 1232 | |||
AK291112.1 | - | 3 | 460 | |||
BC000814.1 | - | 3 | 392 | 3453337 | ||
BC031268.1 | - | 3 | 998 | 5284849 | ||
BE296707.1 | - | 3 | 388 | |||
BE906361.1 | - | 4 | 545 | |||
BM849453.1 | - | 4 | 562 | |||
BQ437864.1 | - | 3 | 624 | |||
BU657748.1 | - | 3 | 480 | |||
BX952985.1 | - | 3 | 393 | |||
CB161219.1 | - | 3 | 536 | |||
CB996365.1 | - | 4 | 720 | |||
CR450333.1 | - | 3 | 308 | |||
X89750.1 | - | 3 | 619 |