Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL23A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173465.3 | NP_775736.2 | 29 | 2229 | ||
XM_006714933.3 | XP_006714996.1 | 30 | 2037 | |||
XM_006714934.3 | XP_006714997.1 | 29 | 2010 | |||
XM_006714935.3 | XP_006714998.1 | 29 | 2001 | |||
XM_006714936.3 | XP_006714999.1 | 29 | 1974 | |||
XM_011534688.2 | XP_011532990.1 | 29 | 2010 | |||
XM_011534689.2 | XP_011532991.1 | 29 | 1983 | |||
XM_011534690.2 | XP_011532992.1 | 28 | 1983 | |||
XM_011534691.2 | XP_011532993.1 | 26 | 1929 | |||
XM_011534692.2 | XP_011532994.1 | 30 | 3062 | |||
GenBank mRNA | AL137461.1 | - | 12 | 856 | ||
AY158895.1 | - | 29 | 2229 | |||
AY898961.1 | - | 29 | 2229 | |||
BC042428.1 | - | 30 | 2011 | 5165083 |