Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MISP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173481.3 | NP_775752.1 | 5 | 2488 | ||
NR_135168.1 | 4 | 651 | ||||
XM_011527685.2 | XP_011525987.1 | 5 | 2533 | |||
XM_011527686.2 | XP_011525988.1 | 5 | 2626 | |||
GenBank mRNA | BC042125.2 | - | 5 | 2455 | 3848272 | |
BC052236.1 | - | 5 | 2475 | 5431360 | ||
BX648389.1 | - | 5 | 2469 | |||
BX648815.1 | - | 4 | 1676 |