Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MROH2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173489.4 | NP_775760.3 | 37 | 4599 | ||
XM_011513952.1 | XP_011512254.1 | 37 | 4585 | |||
XM_011513953.1 | XP_011512255.1 | 36 | 4523 | |||
GenBank mRNA | AF432068.1 | - | 7 | 964 | ||
AK097562.1 | - | 13 | 1645 | |||
AK126005.1 | - | 22 | 2468 | |||
AK128183.1 | - | 27 | 3065 | |||
AL832763.1 | - | 32 | 3524 | |||
BC044937.1 | - | 26 | 3251 | |||
BC052962.1 | - | 37 | 4560 | 4838071 | ||
CR749346.1 | - | 32 | 3500 |