Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTCHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173495.2 | NP_775766.2 | 3 | 2583 | ||
XM_011545449.2 | XP_011543751.1 | 4 | 2668 | |||
GenBank mRNA | AK054858.1 | - | 3 | 2473 | ||
AK298796.1 | - | 3 | 2586 | |||
BC121061.2 | - | 3 | 2430 | 40118449 | ||
KJ534921.1 | - | 3 | 2575 | |||
KJ535084.1 | - | 4 | 2534 | |||
KJ535090.1 | - | 4 | 2739 | |||
KR270726.1 | - | 2 | 1914 |