Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304804.1 | NP_001291733.1 | 2, 3 | 231 | ||
NM_001304805.1 | NP_001291734.1 | 3, 4 | 380 | |||
NM_001304806.1 | NP_001291735.1 | 2, 3 | 294 | |||
NM_173499.4 | NP_775770.1 | 2, 3 | 449 | |||
XM_011521265.2 | XP_011519567.1 | 2, 3 | 496 | |||
GenBank mRNA | AY489187.1 | - | 2, 3 | 413 | ||
BC033979.2 | - | 2, 3 | 386 | 5295377 | ||
BC066292.1 | - | 2, 3 | 408 | 5271997 | ||
BC112390.1 | - | 1, 2 | 777 | 5168326 | ||
HY024478.1 | - | 2, 3 | 257 | |||
HY034703.1 | - | 3, 4 | 377 | |||
HY047940.1 | - | 2, 3 | 231 |