Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258290.1 | NP_001245219.1 | 7 | 838 | ||
NM_001258291.1 | NP_001245220.1 | 7 | 838 | |||
NM_173502.4 | NP_775773.2 | 7 | 838 | |||
XM_011545758.2 | XP_011544060.1 | 6 | 1056 | |||
XM_017022968.1 | XP_016878457.1 | 7 | 827 | |||
XM_017022969.1 | XP_016878458.1 | 7 | 827 | |||
XM_017022970.1 | XP_016878459.1 | 7 | 827 | |||
XM_017022971.1 | XP_016878460.1 | 7 | 827 | |||
GenBank mRNA | AJ627034.1 | - | 7 | 810 | ||
AK075142.1 | - | 7 | 811 | |||
AK290310.1 | - | 7 | 838 | |||
AK297997.1 | - | 5 | 779 | |||
AK301409.1 | - | 7 | 810 | |||
BC137396.1 | - | 7 | 805 | 9021394 | ||
BC144615.1 | - | 7 | 805 | 9053146 |