Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173508.3 | NP_775779.1 | 2 | 518 | ||
GenBank mRNA | AK095031.1 | - | 2 | 299 |