Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC25A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001789.2 | NP_001780.2 | 2 | 564 | ||
NM_201567.1 | NP_963861.1 | 2 | 564 | |||
XM_006713434.2 | XP_006713497.1 | 2 | 659 | |||
XM_006713435.2 | XP_006713498.1 | 2 | 659 | |||
XM_011534316.1 | XP_011532618.1 | 3 | 757 | |||
GenBank mRNA | AF277722.1 | - | 2 | 92 | ||
AK290554.1 | - | 2 | 239 | |||
AY137580.1 | - | 2 | 200 | |||
BC007401.2 | - | 2 | 564 | 2989279 | ||
BC018642.2 | - | 2 | 564 | 3687561 | ||
EU812118.1 | - | 2 | 200 | |||
FJ866789.1 | - | 2 | 200 | |||
M81933.1 | - | 2 | 656 |