Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC25C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287582.1 | NP_001274511.1 | 4 | 632 | ||
NM_001287583.1 | NP_001274512.1 | 4 | 749 | |||
NM_001318098.1 | NP_001305027.1 | 4 | 753 | |||
NM_001790.4 | NP_001781.2 | 4 | 636 | |||
NM_022809.3 | NP_073720.1 | 3 | 541 | |||
XM_005272145.3 | XP_005272202.1 | 4 | 749 | |||
XM_006714739.3 | XP_006714802.1 | 3 | 347 | |||
XM_011543759.2 | XP_011542061.1 | 5 | 420 | |||
XM_011543760.2 | XP_011542062.1 | 4 | 749 | |||
XM_011543761.2 | XP_011542063.1 | 3 | 654 | |||
XM_011543762.1 | XP_011542064.1 | 3 | 454 | |||
XM_011543763.1 | XP_011542065.1 | 2 | 45 | |||
XM_011543764.1 | XP_011542066.1 | 3 | 222 | |||
XM_017010101.1 | XP_016865590.1 | 4 | 627 | |||
GenBank mRNA | AF086323.1 | - | 1 | 383 | ||
AF277723.1 | - | 3 | 222 | |||
AF277724.1 | - | 3 | 222 | |||
AF277725.1 | - | 2 | 127 | |||
AF277726.1 | - | 2 | 127 | |||
AJ304504.1 | - | 2 | 220 | |||
AK097710.1 | - | 4 | 749 | |||
AK301828.1 | - | 5 | 699 | |||
AU125025.1 | - | 4 | 557 | |||
BC019089.2 | - | 4 | 526 | 4858949 | ||
M34065.1 | - | 4 | 525 |