Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF596 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042415.2 | NP_001035880.1 | 4, 5 | 468 | ||
NM_001042416.2 | NP_001035881.1 | 4, 5 | 601 | |||
NM_001287254.1 | NP_001274183.1 | 4, 5 | 459 | |||
NM_001287255.1 | NP_001274184.1 | 4, 5 | 434 | |||
NM_001287256.1 | NP_001274185.1 | 4, 5 | 430 | |||
NM_001287399.1 | NP_001274328.1 | 4, 5 | 519 | |||
NM_173539.3 | NP_775810.2 | 4, 5 | 519 | |||
XM_011534744.1 | XP_011533046.1 | 4, 5 | 441 | |||
XM_017013166.1 | XP_016868655.1 | 3, 4 | 2158 | |||
GenBank mRNA | AK313454.1 | - | 4, 5 | 342 | ||
BC026190.1 | - | 4, 5 | 342 | 4837032 | ||
BG771504.1 | - | 4, 5 | 342 | |||
BM478564.1 | - | 4, 5 | 332 | |||
DA504426.1 | - | 4, 5 | 468 | |||
DB034950.1 | - | 4, 5 | 435 | |||
DB047692.1 | - | 4, 5 | 351 | |||
DB053699.1 | - | 4, 5 | 520 | |||
DB055646.1 | - | 4, 5 | 399 | |||
DN995476.1 | - | 4, 5 | 463 | |||
DR006332.1 | - | 4, 5 | 459 |