Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173558.3 | NP_775829.2 | 2 | 391 | ||
XM_011514372.2 | XP_011512674.1 | 2 | 383 | |||
XM_011514373.2 | XP_011512675.1 | 2 | 383 | |||
XM_017010429.1 | XP_016865918.1 | 2 | 383 | |||
GenBank mRNA | AK024456.1 | - | 1 | 3286 | ||
AK092732.1 | - | 2 | 339 | |||
AK290419.1 | - | 2 | 384 | |||
AK300964.1 | - | 2 | 339 | |||
AK301156.1 | - | 2 | 383 | |||
BC023645.2 | - | 2 | 391 | 5087501 | ||
BC053655.1 | - | 2 | 336 | 5755893 | ||
BX648164.1 | - | 2 | 298 |