Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR14L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173566.2 | NP_775837.2 | 4 | 5817 | ||
XM_006724219.3 | XP_006724282.1 | 4 | 6377 | |||
XM_006724220.3 | XP_006724283.1 | 4 | 5779 | |||
XM_006724221.2 | XP_006724284.1 | 3 | 5333 | |||
XM_011530080.2 | XP_011528382.1 | 5 | 6585 | |||
XM_011530081.2 | XP_011528383.1 | 4 | 6461 | |||
XM_017028736.1 | XP_016884225.1 | 4 | 7537 | |||
XM_017028737.1 | XP_016884226.1 | 3 | 6975 | |||
GenBank mRNA | AK130944.1 | - | 1 | 1631 | ||
BC040859.1 | - | 1 | 531 | 5535580 | ||
BX647272.1 | - | 1 | 120 | |||
CT841517.1 | - | 3 | 5649 |