Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLB1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080407.2 | NP_001073876.2 | 12 | 1490 | ||
XM_011542566.2 | XP_011540868.1 | 11 | 1560 | |||
XM_011542567.2 | XP_011540869.1 | 11 | 1560 | |||
XM_011542568.2 | XP_011540870.1 | 11 | 1524 | |||
XM_011542571.2 | XP_011540873.1 | 11 | 1560 | |||
XM_011542572.1 | XP_011540874.1 | 7 | 708 | |||
XM_011542573.1 | XP_011540875.1 | 5 | 644 | |||
XM_011542574.2 | XP_011540876.1 | 5 | 767 | |||
XM_017017155.1 | XP_016872644.1 | 12 | 2388 | |||
GenBank mRNA | AK074712.1 | - | 3 | 196 | ||
AK124514.1 | - | 1 | 898 | |||
BC131702.1 | - | 3 | 186 | |||
CX760372.1 | - | 6 | 452 |