Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173591.3 | NP_775862.3 | 53 | 6323 | ||
XM_005268802.3 | XP_005268859.1 | 57 | 7124 | |||
XM_011538191.1 | XP_011536493.1 | 54 | 6400 | |||
XM_011538192.2 | XP_011536494.1 | 51 | 6341 | |||
XM_011538193.1 | XP_011536495.1 | 51 | 6260 | |||
GenBank mRNA | AK096852.1 | - | 7 | 601 | ||
BC101016.2 | - | 8 | 558 | |||
BC101017.2 | - | 7 | 472 | |||
BC101018.1 | - | 7 | 472 |