Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135195.1 | NP_001128667.1 | 2 | 338 | ||
NM_173596.2 | NP_775867.2 | 4 | 367 | |||
XM_005268803.1 | XP_005268860.1 | 2 | 255 | |||
XM_011538198.1 | XP_011536500.1 | 2 | 320 | |||
XM_011538199.1 | XP_011536501.1 | 4 | 374 | |||
XM_011538200.1 | XP_011536502.1 | 3 | 324 | |||
XM_011538201.1 | XP_011536503.1 | 3 | 298 | |||
XM_017019185.1 | XP_016874674.1 | 2 | 298 | |||
XM_017019186.1 | XP_016874675.1 | 4 | 485 | |||
XM_017019187.1 | XP_016874676.1 | 2 | 234 | |||
GenBank mRNA | AK172768.1 | - | 2 | 301 | ||
AK313188.1 | - | 4 | 330 | |||
BC027884.1 | - | 2 | 212 | 5221329 | ||
DA640935.1 | - | 2 | 338 | |||
DA918133.1 | - | 3 | 325 |