Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNLDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271862.1 | NP_001258791.1 | 4, 5 | 307 | ||
NM_173516.2 | NP_775787.1 | 4, 5 | 406 | |||
XM_011535491.2 | XP_011533793.1 | 4, 5 | 333 | |||
XM_017010312.1 | XP_016865801.1 | 4, 5 | 1095 | |||
XM_017010314.1 | XP_016865803.1 | 4, 5 | 1095 | |||
GenBank mRNA | AK093139.1 | - | 4, 5 | 284 | ||
AK097559.1 | - | 4, 5 | 386 | |||
BC112246.1 | - | 4, 5 | 290 | 8327714 |