Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173521.4 | NP_775792.4 | 3 | 254 | ||
NR_109816.1 | 5 | 456 | ||||
XM_006716974.2 | XP_006717037.1 | 5 | 440 | |||
XM_006716975.2 | XP_006717038.1 | 5 | 440 | |||
XM_011518302.2 | XP_011516604.1 | 3 | 2249 | |||
XM_011518303.1 | XP_011516605.1 | 3 | 221 | |||
XM_011518309.1 | XP_011516611.1 | 5 | 440 | |||
XM_017014341.1 | XP_016869830.1 | 5 | 440 | |||
GenBank mRNA | AK057341.1 | - | 5 | 391 | ||
AL704322.1 | - | 5 | 457 | |||
AL833535.1 | - | 3 | 249 | |||
BC112357.1 | - | 5 | 384 | |||
BC144680.1 | - | 5 | 380 | |||
DB066382.1 | - | 3 | 254 |